Learn More
1,3,5-Trithiane, TRC
Product Code. 30452310
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452310 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30452310 Supplier Toronto Research Chemicals Supplier No. TRCT88644010G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3,5-Trithiane
CAS 291-21-4
Molecular Formula C3 H6 S3
Quantity 10 g
Synonym s-Trithiane, 1,3,5-Trithiacyclohexane, NSC 1937, Thioform, Thioformaldehyde Trimer, Trimethylene Trisulfide, Trithioformaldehyde
InChI Formula InChI=1S/C3H6S3/c1-4-2-6-3-5-1/h1-3H2
SMILES C1SCSCS1
IUPAC Name 1,3,5-trithiane
Molecular Weight (g/mol) 138.27
Formula Weight 137.9632