missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propanedithiol, 97%

Codice prodotto. 11472308
Cambia vista
Click to view available options
Quantità:
5 g
25 g
100 g
Dimensione della confezione:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11472308 25 g 25g
11462308 5 g 5g
11482308 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11472308 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A15261.14
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 109-80-8 | C3H8S2 | 108.217 g/mol

1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propanedithiol is used as a reagent in the preparation of thioketals and thioacetals. It acts as a flavoring agent. It is used as a precursor in the synthesis of cyclic dithioacetal (1,3-dithiane) derivatives of carbonyl compounds. It is involved in the preparation of diiron propanedithiolate hexacarbonyl by reacting reaction with triiron dodecacarbonyl. Further, it is used for the protection of aldehydes and ketones through their reversible formation of dithianes. In addition to this, it reacts with metal ions to form chelate rings.

Solubility
Slightly soluble in water. Miscible with alcohol, ether, chloroform, etherMiscible with alcohol, ether, chloroform and ether. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 109-80-8
Formula molecolare C3H8S2
Molecular Weight (g/mol) 108.217
Numero MDL MFCD00004904
InChI Key ZJLMKPKYJBQJNH-UHFFFAOYSA-N
Sinonimo 1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol
PubChem CID 8013
ChEBI CHEBI:44864
IUPAC Name propane-1,3-dithiol
SMILES C(CS)CS

Specifica

Materiale o nome chimico 1,3-Propanedithiol
Punto di fusione -79°C
Densità 1.077
Punti di ebollizione 168°C to 170°C
Punto d'infiammabilità 63°C (145°F)
Odore Stench
Indice di rifrazione 1.54
Quantità 25 g
Numero UN UN3334
Beilstein 1071197
Indice di Merck 14,7801
Informazioni di solubilità Slightly soluble in water; Miscible with alcohol,ether,chloroform,etherMiscible with alcohol,ether,chloroform and ether; Slightly miscible with water.
Peso formulazione 108.23
Percent Purity 97%
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 1,3-Propanedithiol
Avvertenza
  • Pericolo
Categoria di pericolo
  • Tossicità acuta Categoria 3
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Liquido infiammabile Categoria 3
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
  • H226-Liquido e vapori infiammabili.
  • H301-Tossico se ingerito.
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H335-Può irritare le vie respiratorie.
Consigli di prudenza
  • P210-Tenere lontano da fonti di calore, superfici riscaldate, scintille, fiamme e altre fonti di innesco. Vietato fumare.
  • P264-Lavare accuratamente dopo l'uso.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P301+P310-IN CASO DI INGESTIONE: contattare immediatamente un CENTRO ANTIVELENI/un medico
  • P303+P361+P353-IN CASO DI CONTATTO CON LA PELLE (o con i capelli): togliere immediatamente tutti gli indumenti contaminati. Sciacquare la pelle/fare una doccia.
  • P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
  • P332+P313-In caso di irritazione della pelle: consultare un medico.
  • P337+P313-Se l'irritazione degli occhi persiste, consultare un medico.
Informazioni supplementari
  • MIXTURE LIST-Contain: 1,3-Propanedithiol

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.