Learn More
1,3-Pyrenediamine, TRC
Specifica
Specifica
| Chemical Name or Material | 1,3-Pyrenediamine |
| CAS | 92821-64-2 |
| Molecular Formula | C16H12N2 |
| Quantity | 25 mg |
| Synonym | 1,3-Diaminopyrene |
| InChI Formula | InChI=1S/C16H12N2/c17-13-8-14(18)12-7-5-10-3-1-2-9-4-6-11(13)16(12)15(9)10/h1-8H,17-18H2 |
| SMILES | Nc1cc(N)c2ccc3cccc4ccc1c2c34 |
| IUPAC Name | pyrene-1,3-diamine |
| Molecular Weight (g/mol) | 232.28 |
| Formula Weight | 232.1 |