Learn More
1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one, Mikromol™
Description
1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one
Specifications
Specifications
| Materiale o nome chimico | 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one |
| CAS | 865792-18-3 |
| Nomi componenti o analita | 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one |
| Shipping Condition | Room Temperature |
| Formula molecolare | C31 H45 N9 O2 |
| Certificazione/Conformità | ISO 9001 |
| Sinonimo | 2(1H)-Quinolinone, 6-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyl-1H-tetrazol-5-yl)butyl]-3,4-dihydro-2(1H)-quinolinone, 1-(4-(5-Cyclohexyl-1H-tetrazol-1-yl)butyl)-6-(4-(1-cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one, Cilostazol Related Compound C |
| InChI Formula | InChI=1S/C31H45N9O2/c41-31-20-17-24-23-27(42-22-10-8-16-30-33-35-37-40(30)26-13-5-2-6-14-26)18-19-28(24)38(31)21-9-7-15-29-32-34-36-39(29)25-11-3-1-4-12-25/h18-19,23,25-26H,1-17,20-22H2 |
| SMILES | O=C1CCc2cc(OCCCCc3nnnn3C4CCCCC4)ccc2N1CCCCc5nnnn5C6CCCCC6 |
| IUPAC Name | 6-[4-(1-cyclohexyltetrazol-5-yl)butoxy]-1-[4-(1-cyclohexyltetrazol-5-yl)butyl]-3,4-dihydroquinolin-2-one |
| Show More |