Learn More
1,4-Diformylpiperazine, 98+%, Thermo Scientific™
Product Code. 11449246
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449246 500 g 500g
11429246 25 g 25g
11439246 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11449246 Supplier Thermo Scientific Alfa Aesar Supplier No. A18097.36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4164-39-0
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00006153
InChI Key CBLGQEBXWDKYDI-UHFFFAOYSA-N
Synonym 1,4-diformylpiperazine, 1,4-piperazinedicarboxaldehyde, pubchem15032, acmc-1ago2, 1,4-piperazine dialdehyde, piperazine-1,4-dicarboxaldehyde, 1,4 piperazinedicarboxaldehyde
PubChem CID 77821
IUPAC Name piperazine-1,4-dicarbaldehyde
SMILES O=CN1CCN(CC1)C=O
Specifications
Melting Point 126°C to 129°C
Boiling Point 154°C (0.25 mmHg)
Odor Odorless
Quantity 500 g
Beilstein 121175
Sensitivity Air Sensitive
Formula Weight 142.16
Percent Purity ≥98%
Chemical Name or Material 1,4-Diformylpiperazine