missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenoxybenzene, 97%, Thermo Scientific Chemicals

Catalog Number p-7022208
50.40 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
5 g
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3061-36-7
C18H14O2
262.308
MFCD00038368
UVGPELGZPWDPFP-UHFFFAOYSA-N
benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
520487
CHEBI:39271
1,4-diphenoxybenzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3061-36-7
C18H14O2
262.308
MFCD00038368
UVGPELGZPWDPFP-UHFFFAOYSA-N
benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
520487
CHEBI:39271
1,4-diphenoxybenzene
C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1,4-Diphenoxybenzene
Punto di fusione 72°C to 73°C
Densità 1.083
Punti di ebollizione 172°C (0.01 mmHg)
Punto d'infiammabilità 202°C (395°F)
Quantità 5 g
Beilstein 2215945
Peso formulazione 262.31
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'