Learn More
Trajan Scientific and Medical™ 1/4 in. Graphite Fittings
Descrizione
- Easy to use
- Forms reliable seal
- Reusable
- Soft material, forms soft grip to the capillary column
- Low emissions
Specifica
Specifica
| Tipo | Graphite Reducing Ferrule 1/4 in. Fitting 5/16 in. x 9.5mm x 4.6mm |
| Dimensioni sistema di montaggio (sistema metrico) | 1/4 in. |
| Dimensioni (sistema metrico) | 9.5 mm x 4.6 mm |
| Materiale | Graphite |
| Quantità | 10/Pk. |
| Diameter (Metric) Inside | 5/16 in. |