Learn More
1-(4-Isopropylphenyl)ethanone (4-Isopropylacetophenone), Mikromol™
Product Code. 30051973
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051973 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051973 Supplier Mikromol Supplier No. MM0002.57
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(4-Isopropylphenyl)ethanone (4-Isopropylacetophenone)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(4-Isopropylphenyl)ethanone (4-Isopropylacetophenone)
CAS 645-13-6
Analyte or Component Names 1-(4-Isopropylphenyl)ethanone (4-Isopropylacetophenone)
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H14 O
Certifications/Compliance ISO 17025
Synonym 1-(4-Isopropylphenyl)ethanone, 4-Isopropylacetophenone
InChI Formula InChI=1S/C11H14O/c1-8(2)10-4-6-11(7-5-10)9(3)12/h4-8H,1-3H3
SMILES CC(C)c1ccc(cc1)C(=O)C
IUPAC Name 1-(4-propan-2-ylphenyl)ethanone
Molecular Weight (g/mol) 162.23
Quantity 100 mg
Formula Weight 162.1045 g/mol