Learn More
1-(4-Trifluoromethylphenyl)pentan-1-one, TRC
Product Code. 30455553
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455553 2.5 g 2.5g
30451783 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30455553 Supplier Toronto Research Chemicals Supplier No. TRCT7914202.5G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42916-66-5
Molecular Formula C12H13F3O
Molecular Weight (g/mol) 230.23
Synonym 1-[4-(Trifluoromethyl)phenyl]-1-pentanone, p-(Trifluoromethyl)valerophenone, 1-(4-Trifluoromethylphenyl)pentan-1-one
IUPAC Name 1-(4-(trifluoromethyl)phenyl)pentan-1-one
SMILES O=C(CCCC)C1=CC=C(C(F)(F)F)C=C1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C12H13F3O/c1-2-3-4-11(16)9-5-7-10(8-6-9)12(13,14)15/h5-8H,2-4H2,1H3
Formula Weight 230.09
Chemical Name or Material 1-(4-Trifluoromethylphenyl)pentan-1-one