missing translation for 'onlineSavingsMsg'
Learn More

1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals

Catalog Number p-7023309
Click to view available options
Quantità:
0.25 g
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
498-07-7
C6H10O5
162.14
MFCD00063248
TWNIBLMWSKIRAT-UHFFFAOYNA-N
1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucose
2724705
CHEBI:30997
(1R,2S,3S,4R,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
OC1C2COC(O2)C(O)C1O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
498-07-7
C6H10O5
162.14
MFCD00063248
TWNIBLMWSKIRAT-UHFFFAOYNA-N
1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucose
2724705
CHEBI:30997
(1R,2S,3S,4R,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
OC1C2COC(O2)C(O)C1O

1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

Solubility
Soluble in Ethanol and Water.

Notes
Store at -20°C. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1,6-Anhydro-beta-D-glucopyranose
Punto di fusione 180°C to 184°C
Quantità 0.25 g
Beilstein 80998
Informazioni di solubilità Soluble in Ethanol and Water.
Peso formulazione 162.14
Percent Purity 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'