Learn More
1,8-Dioxacyclotetradecane-2,9-dione, TRC
Product Code. 30376711
Change view
Click to view available options
Quantità:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30376711 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30376711 Supplier Toronto Research Chemicals Supplier No. TRCD426815100MG
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Materiale o nome chimico 1,8-Dioxacyclotetradecane-2,9-dione
CAS 1020-83-3
Formula molecolare C12 H20 O4
Quantità 100 mg
Sinonimo 2-Oxepanone, dimer, Caprolactone cyclic dimer, Dicaprolactone, ε-Caprolactone cyclic dimer
InChI Formula InChI=1S/C12H20O4/c13-11-7-3-1-5-9-15-12(14)8-4-2-6-10-16-11/h1-10H2
SMILES O=C1CCCCCOC(=O)CCCCCO1
IUPAC Name 1,8-dioxacyclotetradecane-2,9-dione
Molecular Weight (g/mol) 228.285
Peso formulazione 228.136