Learn More
1-Acenaphthenone, TRC
Product Code. 30310964
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310964 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310964 Supplier Toronto Research Chemicals Supplier No. TRCA130755250MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Acenaphthenone
CAS 2235-15-6
Molecular Formula C12 H8 O
Quantity 250 mg
Synonym 1-Acenaphthone, 2-Acenaphthone, Acenaphthenone, Acenaphthylen-1-one, NSC 18756, NSC 22341, 1-Acenaphthenone (6CI,7CI,8CI), 1,2-Dihydroacenaphthylen-1-one, 2H-Acenaphthylen-1-one
InChI Formula InChI=1S/C12H8O/c13-11-7-9-5-1-3-8-4-2-6-10(11)12(8)9/h1-6H,7H2
SMILES O=C1Cc2cccc3cccc1c23
IUPAC Name 2H-acenaphthylen-1-one
Molecular Weight (g/mol) 168.19
Formula Weight 168.0575