Learn More
1-Acenaphthenone, TRC
Product Code. 30319137
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30319137 Supplier Toronto Research Chemicals Supplier No. TRCA1307555G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Acenaphthenone
CAS 2235-15-6
Molecular Formula C12 H8 O
Quantity 5 g
Synonym 1-Acenaphthone, 2-Acenaphthone, Acenaphthenone, Acenaphthylen-1-one, NSC 18756, NSC 22341, 1-Acenaphthenone (6CI,7CI,8CI), 1,2-Dihydroacenaphthylen-1-one, 2H-Acenaphthylen-1-one
InChI Formula InChI=1S/C12H8O/c13-11-7-9-5-1-3-8-4-2-6-10(11)12(8)9/h1-6H,7H2
SMILES O=C1Cc2cccc3cccc1c23
IUPAC Name 2H-acenaphthylen-1-one
Molecular Weight (g/mol) 168.19
Formula Weight 168.0575