Learn More
1-Allylimidazole, 97%
Product Code. 11397516
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397516 50 g 50g
11387516 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11397516 Supplier Thermo Scientific Alfa Aesar Supplier No. L11053.18
 Restricted Item
Spedizione stimata: 01-12-2026
Add to basket
CAS: 31410-01-2 | C6H8N2 | 108.14 g/mol
1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Allylimidazole, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 31410-01-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00037894
InChI Key XLXCHZCQTCBUOX-UHFFFAOYSA-N
Synonym 1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #
PubChem CID 35794
IUPAC Name 1-prop-2-enylimidazole
SMILES C=CCN1C=CN=C1
Spezifikation
Density 1.003
Boiling Point 212°C to 213°C
Flash Point 101°C (213°F)
Refractive Index 1.505
Quantity 50 g
UN Number UN3267
Beilstein 605953
Solubility Information It is soluble in water.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 1-Allylimidazole
Mehr anzeigen Weniger anzeigen
RUO – Research Use Only
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.