Learn More
1-Aminocyclohexanecarboxylic acid, 98%
Codice prodotto. 11478980
Click to view available options
Quantità:
5 g
25 g
Dimensione della confezione:
25g
5g
Codice prodotto. 11478980
Marca: Thermo Scientific Alfa Aesar A11853.14
Richiesta di formato massivo o personalizzato
CAS: 2756-85-6 | C7H13NO2 | 143.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 2756-85-6
Formula molecolare C7H13NO2
Molecular Weight (g/mol) 143.19
Numero MDL MFCD00001487
InChI Key WOXWUZCRWJWTRT-UHFFFAOYSA-N
Sinonimo 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd
PubChem CID 1366
ChEBI CHEBI:86534
IUPAC Name 1-aminocyclohexane-1-carboxylic acid
SMILES NC1(CCCCC1)C(O)=O
Specifica
Materiale o nome chimico 1-Aminocyclohexanecarboxylic acid
Punto di fusione >300°C
Quantità 25 g
Beilstein 2355692
Sensibilità Hygroscopic
Peso formulazione 143.19
Percent Purity 98%