Learn More
1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid, 96%
Product Code. 11424510
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424510 1 g 1g
11414510 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11424510 Supplier Thermo Scientific Alfa Aesar Supplier No. H50086.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 1210863-93-6 | C12H19NO6 | 273.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1210863-93-6
Molecular Formula C12H19NO6
Molecular Weight (g/mol) 273.29
MDL Number MFCD07781134
InChI Key OHQXZMNJROMWMW-UHFFFAOYNA-N
Synonym 4-acetoxy-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid, 1-boc-4-methoxycarbonyl pyrrolidine-3-carboxylic acid, pyrrolidine-1,3,4-tricarboxylic acid 1-tert-butyl 3-methyl diester, 4-acetyloxy-1-tert-butoxycarbonyl pyrrolidine-3-carboxylic acid
PubChem CID 45925717
IUPAC Name 4-acetyloxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-3-carboxylic acid
SMILES CC(=O)OC1CN(CC1C(=O)O)C(=O)OC(C)(C)C
Specifications
Quantity 1 g
Formula Weight 273.29
Percent Purity 96%
Chemical Name or Material 1-Boc-4-(methoxycarbonyl)pyrrolidine-3-carboxylic acid