Learn More
1-Bromo-2-naphthaldehyde, TRC
Product Code. 30330650
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330650 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30330650 Supplier Toronto Research Chemicals Supplier No. TRCB6884551G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromo-2-naphthaldehyde
CAS 3378-82-3
Molecular Formula C11H7BrO
Quantity 1 g
Synonym 1-Bromo-2-naphthalenecarboxaldehyde
InChI Formula InChI=1S/C11H7BrO/c12-11-9(7-13)6-5-8-3-1-2-4-10(8)11/h1-7H
SMILES O=CC1=CC=C2C=CC=CC2=C1Br
IUPAC Name 1-bromo-2-naphthaldehyde
Molecular Weight (g/mol) 235.08
Formula Weight 233.97