missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dimethyladamantane, Mikromol™

Codice prodotto. 17959024
Cambia vista
Click to view available options
Quantità:
25 mg
Dimensione della confezione:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17959024 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17959024 Fornitore Mikromol N. del fornitore MM1045.050025
 Articolo soggetto a restrizioni
Spedizione stimata: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Bromo-3,5-dimethyladamantane
TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 1-Bromo-3,5-dimethyladamantane
CAS 941-37-7
Nomi componenti o analita 1-Bromo-3,5-dimethyladamantane
Shipping Condition Room Temperature
Formula molecolare C12 H19 Br
Certificazione/Conformità ISO 17025
Sinonimo 1-Bromo-3,5-dimethyladamantane, Memantine Related Compound D (USP), Memantine Related Compound D
InChI Formula InChI=1S/C12H19Br/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9H,3-8H2,1-2H3
SMILES CC12CC3CC(C)(C1)CC(Br)(C3)C2
IUPAC Name 1-bromo-3,5-dimethyladamantane
Molecular Weight (g/mol) 243.18
Quantità 25 mg
Peso formulazione 242.067 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.