missing translation for 'onlineSavingsMsg'
Learn More

((1-Chloro-2-methylpropan-2-yl)oxy)trimethylsilane, TRC

Product Code. 30352291
Change view
Click to view available options
Quantità:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30352291 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352291 Supplier Toronto Research Chemicals Supplier No. TRCC8975452.5G

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico ((1-Chloro-2-methylpropan-2-yl)oxy)trimethylsilane
CAS 31557-15-0
Formula molecolare C7H17ClOSi
Quantità 2.5 g
InChI Formula InChI=1S/C7H17ClOSi/c1-7(2,6-8)9-10(3,4)5/h6H2,1-5H3
SMILES CC(C)(CCl)O[Si](C)(C)C
IUPAC Name (1-chloro-2-methylpropan-2-yl)oxy-trimethylsilane
Molecular Weight (g/mol) 180.75
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.