Learn More
1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | 1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester |
| CAS | 94242-51-0 |
| Formula molecolare | C15 H12 F3 N O3 |
| Quantità | 10 mg |
| Sinonimo | 3-Quinolinecarboxylic acid, 1-cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-, ethyl ester, Ethyl 1-cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylate, 1-Cyclopropyl-1,4-dihydro-6,7,8-trifluoro-4-oxoquinoline-3-carboxylic Acid Ethyl Ester, 1-Cyclopropyl-6,7,8-trifluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C15H12F3NO3/c1-2-22-15(21)9-6-19(7-3-4-7)13-8(14(9)20)5-10(16)11(17)12(13)18/h5-7H,2-4H2,1H3 |
| SMILES | CCOC(=O)C1=CN(C2CC2)c3c(F)c(F)c(F)cc3C1=O |
| IUPAC Name | ethyl 1-cyclopropyl-6,7,8-trifluoro-4-oxoquinoline-3-carboxylate |
| Molecular Weight (g/mol) | 311.2559 |
| Peso formulazione | 311.0769 |