Learn More
1-Ethynylpyrene, 96%, Thermo Scientific Chemicals
Click to view available options
Quantità:
1 g
5 g
Descrizione
1-Ethynylpyrene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethynylpyrene is used as a pharmaceutical intermediate.
Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
1-Ethynylpyrene is used as a pharmaceutical intermediate.
Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifica
Materiale o nome chimico | 1-Ethynylpyrene |
Punto di fusione | 113°C to 116°C |
Quantità | 1 g |
Beilstein | 2365008 |
Informazioni di solubilità | Soluble in chloroform,dichloromethane and toluene. Insoluble in water. |
Peso formulazione | 226.28 |
Percent Purity | 96% |