missing translation for 'onlineSavingsMsg'
Learn More

1-Hexen-3-ol, 98%, Thermo Scientific Chemicals

Catalog Number 11404217
Click to view available options
Quantità:
10 g
50 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
4798-44-1
C6H12O
100.161
MFCD00004581
BVOSSZSHBZQJOI-UHFFFAOYSA-N
1-hexen-3-ol, propylvinylcarbinol, 3-hydroxy-1-hexene, propyl vinyl carbinol, vinyl propyl carbinol, 1-vinylbutanol, 1-vinylbutan-1-ol, fema no. 3608, 1-hexene-3-ol, 1-hexen-3-ol, r
20928
hex-1-en-3-ol
CCCC(C=C)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
4798-44-1
C6H12O
100.161
MFCD00004581
BVOSSZSHBZQJOI-UHFFFAOYSA-N
1-hexen-3-ol, propylvinylcarbinol, 3-hydroxy-1-hexene, propyl vinyl carbinol, vinyl propyl carbinol, 1-vinylbutanol, 1-vinylbutan-1-ol, fema no. 3608, 1-hexene-3-ol, 1-hexen-3-ol, r
20928
hex-1-en-3-ol
CCCC(C=C)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1-Hexen-3-ol
Densità 0.837
Punti di ebollizione 132°C to 134°C
Punto d'infiammabilità 35°C (95°F)
Odore Characteristic
Indice di rifrazione 1.429
Quantità 50 g
Numero UN UN1987
Beilstein 1720166
Peso formulazione 100.16
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'