missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropyl-3-(4-fluorophenyl)indole-2-carboxaldehyde, TRC

Product Code. 30406389
Change view
Click to view available options
Quantità:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30406389 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406389 Supplier Toronto Research Chemicals Supplier No. TRCI8245302.5G
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 1-Isopropyl-3-(4-fluorophenyl)indole-2-carboxaldehyde
CAS 101125-34-2
Formula molecolare C18 H16 F N O
Quantità 2.5 g
Sinonimo 1H-Indole-2-carboxaldehyde, 3-(4-fluorophenyl)-1-(1-methylethyl)-, 3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indole-2-carboxaldehyde, 3-(4-Fluorophenyl)-1-isopropyl-1H-indole-2-carbaldehyde, 3-(4-Fluorophenyl)-1-isopropyl-1H-indole-2-carboxaldehyde
InChI Formula InChI=1S/C18H16FNO/c1-12(2)20-16-6-4-3-5-15(16)18(17(20)11-21)13-7-9-14(19)10-8-13/h3-12H,1-2H3
SMILES CC(C)n1c(C=O)c(c2ccc(F)cc2)c3ccccc13
IUPAC Name 3-(4-fluorophenyl)-1-propan-2-ylindole-2-carbaldehyde
Molecular Weight (g/mol) 281.32
Peso formulazione 281.1216
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.