Learn More
1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%
Product Code. 15499005
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499005 25 g 25g
15489005 5 g 5g
15409015 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15499005 Supplier Thermo Scientific Alfa Aesar Supplier No. H35352.14
 Restricted Item
In stock
Add to basket
CAS: 178631-04-4 | C14H23F6N3O4S2 | 475.465 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 178631-04-4
Molecular Formula C14H23F6N3O4S2
Molecular Weight (g/mol) 475.465
MDL Number MFCD08458946
InChI Key LECQXINNQGHJBM-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide
PubChem CID 12184591
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium
SMILES CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Specifications
Quantity 25 g
Formula Weight 475.46
Percent Purity 99%
Chemical Name or Material 1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide