Learn More
1-Nonanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, ACROS Organics™
Marca: Acros Organics 396700050
Dettagli aggiuntivi : CAS : 35192-74-6 Peso : 0.05500kg
Quantità | 5g |
---|---|
Imballaggio | Glass bottle |
Descrizione
For ion pair chromatographyIdentificatori chimici
35192-74-6 | |
230.3 | |
RUYRDULZOKULPK-UHFFFAOYSA-M | |
23664777 | |
CCCCCCCCCS(=O)(=O)[O-].[Na+] |
C9H19NaO3S | |
MFCD00025010 | |
sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate | |
sodium;nonane-1-sulfonate |
Specifica
0.02 max. (C=0.2 M in water) at 230nm, 0.02 max. (C=0.2 M in water) at 260nm, 0.03 max. (C=0.2 M in water) at 220nm, 0.05 max. (C=0.2 M in water) at 210nm | |
Authentic | |
Glass bottle | |
White | |
5g | |
35192-74-6 | |
100.0 | |
C9H19NaO3S | |
MFCD00025010 | |
Solubility in water: soluble | |
CCCCCCCCCS(=O)(=O)[O-].[Na+] | |
230.3 | |
230.3 | |
99+%, |
1-Nonanesulfonic acid, sodium salt | |
2% max. (150°C) | |
(HPLC, gradient) Passes | |
>300°C | |
Anhydrous | |
99.0 | |
99+% | |
S(O3Na)(CH2)8CH3 | |
sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate | |
RUYRDULZOKULPK-UHFFFAOYSA-M | |
sodium;nonane-1-sulfonate | |
23664777 | |
Crystalline Powder | |
Ion Pair Chromatography |