Learn More
1-Oxo-1H-isochromene-4-carboxylic Acid, TRC
Product Code. 30413352
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413352 10 mg -
30428759 100 mg 100mg
30424959 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30413352 Supplier Toronto Research Chemicals Supplier No. TRCO99165310MG
 Restricted Item
Spedizione stimata: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15868-29-8
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.15
IUPAC Name 1-oxoisochromene-4-carboxylic acid
SMILES OC(=O)C1=COC(=O)c2ccccc12
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C10H6O4/c11-9(12)8-5-14-10(13)7-4-2-1-3-6(7)8/h1-5H,(H,11,12)
Formula Weight 190.0266
Chemical Name or Material 1-Oxo-1H-isochromene-4-carboxylic Acid