Learn More
1-Pentanesulfonic acid, sodium salt hydrate, 99%, Thermo Scientific Chemicals
Catalog Number 10169322
Click to view available options
Quantità:
5 g
25 g
100 g
Imballaggio:
Glass bottle
1266615-65-9
C5H11NaO3S
174.19
MFCD00149548
ROBLTDOHDSGGDT-UHFFFAOYSA-M
n-Amylsulfonic acid, sodium salt
sodium pentane-1-sulfonate
[Na+].CCCCCS([O-])(=O)=O
1266615-65-9
C5H11NaO3S
174.19
MFCD00149548
ROBLTDOHDSGGDT-UHFFFAOYSA-M
n-Amylsulfonic acid, sodium salt
sodium pentane-1-sulfonate
[Na+].CCCCCS([O-])(=O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1-Pentanesulfonic acid, sodium salt hydrate
Punto di fusione >300.0°C
Colore White
Forma fisica Crystalline Powder
Quantità 5 g
Intervallo percentuale saggio 99%
Formula lineare CH3(CH2)4SO3Na·xH2O
Beilstein 04, III, 23
Informazioni di solubilità (10% in water) Clear colorless
Peso formulazione 174.12
Percent Purity 99%
Spettro a infrarossi Authentic
Perdita all'essiccatura 9.6% max. (100°C, 3 hrs) (vacuum dried)
Imballaggio Glass bottle
Aspetto (10% aq. soln.) clear, colorless