missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pyridin-3-yl-piperazine Hydrochloride, TRC
Specifications
Specifications
| Materiale o nome chimico | 1-Pyridin-3-yl-piperazine Hydrochloride |
| CAS | 1010133-97-7 |
| Formula molecolare | C9H13N3•HCl |
| Quantità | 250 mg |
| Sinonimo | 1-(3-Pyridinyl)piperazine Hydrochloride, 1-(3-Pyridyl)piperazine Hydrochloride, 1-(Pyridin-3-yl)piperazine Hydrochloride, 3-(Piperazin-4-yl)pyridine Hydrochloride, NSC 505538 Hydrochloride |
| InChI Formula | InChI=1S/C9H13N3.ClH/c1-2-9(8-11-3-1)12-6-4-10-5-7-12;/h1-3,8,10H,4-7H2;1H |
| SMILES | C1=CC(=CN=C1)N2CCNCC2.Cl |
| IUPAC Name | 1-(pyridin-3-yl)piperazine hydrochloride |
| Molecular Weight (g/mol) | 199.68 |
| Peso formulazione | 199.09 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?