Learn More
Avantor™ J.T. Baker™ SAF-T-SPILL™ NEUTRASORB™ Acid Spill Kit and Components
Be prepared for hazardous laboratory spills. For acid spill clean-up, NEUTRASORB™ provides rapid, controlled neutralization of concentrated acids, and visible color change as indication of complete neutralization.
Marca: Avantor™ 4442
Dettagli aggiuntivi : Peso : 4.10000kg
Descrizione
- Provides rapid, controlled neutralization of concentrated acids, and visible color change as indication of complete neutralization
- Each kit is supplied with the appropriate neutraliser, safety glasses, scoops, gloves, disposal bags and ties, and labels.
- Restock your spill control center (or create your own) with replacement kits, available separately.
- Caution: NEUTRASORB™ should not be used for spills of hydrofluoric acid, fuming nitric acid, fuming sulfuric acid, or peroxy (per-) organic or other highly reactive acids.
Specifica
Purple | |
Granules | |
3.2 kg |
Acid Spill Clean-up Kit | |
Bromocresol Purple Indicator Grade (CAS 115-40-2; < 0.1%), Calcium Carbonate (CAS 471-34-1; 5 - 25%), Magnesium Oxide (CAS 1309-48-4; 25 - 45%), Sodium Carbonate (CAS 497-19-8; 35 - 55%) |