Learn More
Testo™ 108-2 Waterproof Digital Food Thermometer
Descrizione
- Easy to operate and use: lockable probe for a secure hold
- Temperature measuring instrument with connected probe is waterproof (IP67)
- Includes soft case and probe type T
- HACCP-compliant and certified to EN 13485
- Sleek, compact, waterproof housing
- Allows for spot check measurements in seconds, during transportation and storage, in large kitchens or restaurants
- Can be cleaned under running water
- Probe can be locked on the temperature measuring instrument, ensuring a secure hold
- Includes thermometer, lockable type T thermocouple, user guide, calibration protocol and batteries
Specifica
Specifica
| Certificazione/Conformità | HACCP-compliant and certified to EN 13485; Protection Class IP67 with TopSafe; 2004/108/EG |
| Dimensioni (L x P x A) | 140 x 60 x 24 mm |
| Materiale | TPE, ABS, Polycarbonate |
| Tempo di misurazione | 10 sec. reaction time |
| Risoluzione | 0,1°C |
| Peso (sistema metrico) | 154 g (incl. battery, without softcase) |
| Temperature Range (Metric) | -50°C to +300°C |
| Autonomia della batteria | 2500 hr. |
| Tipo di batteria | 3 x AAA micro |
| Accuracy | ±0.5°C from −30° to +70°C |
| Vedi altri risultati |