missing translation for 'onlineSavingsMsg'
Learn More

Furagin-13C3, TRC

Codice prodotto. 30136802
Cambia vista
Click to view available options
Quantità:
1 mg
5 mg
10 mg
Dimensione della confezione:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30136802 10 mg 10mg
30132993 1 mg 1mg
30003894 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30136802 Fornitore Toronto Research Chemicals N. del fornitore TRCF86300210MG

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Formula molecolare C713C3H8N4O5
Molecular Weight (g/mol) 267.17
Sinonimo 1-[[3-(5-Nitro-2-furanyl)-2-propen-1-ylidene]amino]-2,4-imidazolidinedione-13C3, 1-[3-(5-Nitro-2-furyl)allylidenamino]hydantoin-13C3, Akritoin-13C3, F 35-13C3, Furazidin-13C3, Furazidine-13C3, NF 416-13C3;
SMILES O=[13C]1[13C]N(/N=C/C=C/C2=CC=C([N+]([O-])=O)O2)[13C](N1)=O

Specifications

Materiale o nome chimico Furagin-13C3
Quantità 10 mg
InChI Formula InChI=1S/C10H8N4O5/c15-8-6-13(10(16)12-8)11-5-1-2-7-3-4-9(19-7)14(17)18/h1-5H,6H2,(H,12,15,16)/b2-1+,11-5-/i6+1,8+1,10+1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.