missing translation for 'onlineSavingsMsg'
Learn More

11-Aminoundecanoic acid, 97%

Product Code. 10529790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10529790 100 g 100g
10655902 5 g 5g
10284460 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10529790 Supplier Thermo Scientific Acros Supplier No. 104711000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2432-99-7 | C11H23NO2 | 201.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-99-7
Molecular Formula C11H23NO2
Molecular Weight (g/mol) 201.31
MDL Number MFCD00008150
InChI Key GUOSQNAUYHMCRU-UHFFFAOYSA-N
Synonym undecanoic acid, 11-amino, 11-aminoundecylic acid, 11-amino-undecanoic acid, omega-aminoundecanoic acid, ccris 39, unii-sfu34976hb, rilsan t, aminoundecanoicacid, reilsan es natural, 11-amino-undecanoicaci
PubChem CID 17083
ChEBI CHEBI:82387
IUPAC Name 11-aminoundecanoic acid
SMILES C(CCCCCN)CCCCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 187°C to 190°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula H2N(CH2)10CO2H
Quantity 100 g
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in butanol
Formula Weight 201.31
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 11-Aminoundecanoic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.