Learn More
Hamilton™ 13 gauge, Metal Hub Needle, custom length (0.375 to 12 in), point style 5 or AS, 6/PK
Descrizione
Specification Sheet
- Gauge – 13 gauge
- Point Style – 5 or AS
- Needle Length – Custom Needle Length
- Autoclavable – Yes
- Sales Quantity – 6 needles per pack
- Hub – Metal (N)
Specifica
Specifica
| Calibro dell'ago | 13 G |
| Product Type | Metal Hub Needle |