missing translation for 'onlineSavingsMsg'
Learn More

16,17Alpha-Epoxypregnenolone, TRC

Codice prodotto. 30376606
Cambia vista
Click to view available options
Quantità:
100 mg
Dimensione della confezione:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30376606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376606 Supplier Toronto Research Chemicals Supplier No. TRCE592315100MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 16,17alpha-Epoxypregnenolone
CAS 974-23-2
Formula molecolare C21H30O3
Quantità 100 mg
Sinonimo 16alpha,17-Epoxy-3beta-hydroxypregn-5-en-20-one, 16alpha,17alpha-Epoxy-3beta-hydroxy-5-pregnen-20-one, 16alpha,17alpha-Epoxypregn-5-en-3beta-ol-20-one, 16alpha,17alpha-Epoxypregnenolone, 16alpha,17alpha-Oxidopregnenolone, 3beta-Hydroxy-16alpha,17alpha-epoxy-5-pregnen-20-one, 5-Pregnen-16alpha,17alpha-epoxy-3beta-ol-20-one, NSC 122401
InChI Formula InChI=1S/C21H30O3/c1-12(22)21-18(24-21)11-17-15-5-4-13-10-14(23)6-8-19(13,2)16(15)7-9-20(17,21)3/h4,14-18,23H,5-11H2,1-3H3/t14-,15+,16-,17-,18+,19-,20-,21+/m0/s1
SMILES CC(=O)[C@@]12O[C@@H]1C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C
Molecular Weight (g/mol) 330.46
Peso formulazione 330.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.