Learn More
16-Hydroxyhexadecanoic acid, 98%, ACROS Organics™
Product Code. 10761451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10761451 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10761451 Supplier Thermo Scientific Acros Supplier No. 175900010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 506-13-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 95°C to 99°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C16H32O3
Linear Formula HO(CH2)15CO2H
MDL Number MFCD00002750
Quantity 1 g
Beilstein 03, 362
Synonym juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid
InChI Key UGAGPNKCDRTDHP-UHFFFAOYSA-N
SMILES C(CCCCCCCC(=O)O)CCCCCCCO
IUPAC Name 16-hydroxyhexadecanoic acid
Molecular Weight (g/mol) 272.429
PubChem CID 10466
ChEBI CHEBI:55328
Formula Weight 272.42
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 16-Hydroxyhexadecanoic acid