Learn More
Fisherbrand™ Plastic Lock Bacti Caps
Product Code. 11517153 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Green
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11517153 Green Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11517153
Spedizione stimata: 09-12-2026
Add to basket
Add to basket
Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.
  • Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
  • Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm
  • The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur. Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters
  • Improved culture growth
  • Color coded
  • Controlled gas interchange
Order Info
dimClosureColor
Specifications
Product Type Lock Bacti Cap
Autoclavable Autoclavable
Material Plastic
Color Green
Quantity 100 Pack
Size (Metric) 16 mm
For Use With (Equipment) Test Tubes
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.