Learn More
(1RS)-1-Phenyl-1-(pyridin-2-yl)ethanol Hydrochloride, Mikromol™
Descrizione
(1RS)-1-Phenyl-1-(pyridin-2-yl)ethanol Hydrochloride
Specifica
Specifica
| Materiale o nome chimico | (1RS)-1-Phenyl-1-(pyridin-2-yl)ethanol Hydrochloride |
| CAS | 879671-67-7 |
| Nomi componenti o analita | (1RS)-1-Phenyl-1-(pyridin-2-yl)ethanol Hydrochloride |
| Shipping Condition | Room Temperature |
| Formula molecolare | C13 H13 N O . Cl H |
| Certificazione/Conformità | ISO 17025 |
| Sinonimo | Doxylamine Hydrogen Succinate Imp. B (EP), Doxylamine Imp. B (EP), (1RS)-1-Phenyl-1-(pyridin-2-yl)ethanol Hydrochloride, Doxylamine Hydrogen Succinate Imp. B (EP) as Hydrochloride, Doxylamine Hydrogen Succinate Impurity B as Hydrochloride, Doxylamine Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C13H13NO.ClH/c1-13(15,11-7-3-2-4-8-11)12-9-5-6-10-14-12;/h2-10,15H,1H3;1H |
| SMILES | Cl.CC(O)(c1ccccc1)c2ccccn2 |
| IUPAC Name | 1-phenyl-1-pyridin-2-ylethanol;hydrochloride |
| Show More |