missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S,3R,5S)-(+)-2,3-Pinanediol, 98%, Thermo Scientific Chemicals

Catalog Number p-3737227
45.80 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
2.5 g
10 g
100 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
18680-27-8
C10H18O2
170.25
MFCD00077851
MOILFCKRQFQVFS-OORONAJNSA-N
1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s
10219606
(1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
CC1(C2CC1C(C(C2)O)(C)O)C
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
18680-27-8
C10H18O2
170.25
MFCD00077851
MOILFCKRQFQVFS-OORONAJNSA-N
1s,2s,3r,5s-+-2,3-pinanediol, 1s,2s,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptane-2,3-diol, 1s,2s,3r,5s-+-pinanediol, +-pinanediol, pinanediol, +, unii-y6zcv4avra, +-2-hydroxyisopinocampheol, y6zcv4avra, 1s,2s,3r,5s-2,3-pinanediol, bicyclo 3.1.1 heptane-2,3-diol, 2,6,6-trimethyl-, 1s,2s,3r,5s
10219606
(1S,3R,4S,5S)-4,6,6-trimethylbicyclo[3.1.1]heptane-3,4-diol
CC1(C2CC1C(C(C2)O)(C)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico (1S,2S,3R,5S)-(+)-2,3-Pinanediol
CAS min. % 97.5
CAS max % 100.0
Punto di fusione 52°C to 60°C
Colore White to Beige
Punto d'infiammabilità >110°C
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97.5% min. (GC)
Imballaggio Glass bottle
Quantità 2.5 g
Beilstein 06,4145
Condizione di rotazione specifica +8.50 (20.00°C c=6.5,toluene)
Rotazione specifica + 8.5
Peso formulazione 170.25
Percent Purity 98%
Forma fisica Crystalline Powder, Crystals or Crystalline Mass
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'