Learn More
2-(1-Naphthyl)ethanol, TRC
Product Code. 30472790
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472790 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30472790 Supplier Toronto Research Chemicals Supplier No. TRCN3783251G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(1-Naphthyl)ethanol
CAS 773-99-9
Molecular Formula C12 H12 O
Quantity 1 g
InChI Formula InChI=1S/C12H12O/c13-9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7,13H,8-9H2
SMILES OCCc1cccc2ccccc12
IUPAC Name 2-naphthalen-1-ylethanol
Molecular Weight (g/mol) 172.22
Formula Weight 172.0888