missing translation for 'onlineSavingsMsg'
Learn More

2-(2-chlorophenoxy)ethanimidamide hydrochloride, 97%, Thermo Scientific™

Catalog Number p-2696058
Click to view available options
Quantità:
1 g
10 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
58403-03-5
C8H10Cl2N2O
221.08
MFCD00173805
WPWKAEFYVGWTAE-UHFFFAOYSA-N
2-2-chlorophenoxy acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide hydrochloride, 2-2-chlorophenoxy acetimidamide hydrochloride, 2-2-chloro-phenoxy-acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide monohydrochloride, 2-2-chlorophenoxy acetamide hydrochloride, ethanimidamide, 2-2-chlorophenoxy-, monohydrochloride, 2-2-chlorophenoxy ethanamidine, chloride, 2-2-chlorophenoxy acetamidine hcl, 2-chlorophenoxy ethanimidamide hydrochloride
2745914
[H+].[Cl-].NC(=N)COC1=CC=CC=C1Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
58403-03-5
C8H10Cl2N2O
221.08
MFCD00173805
WPWKAEFYVGWTAE-UHFFFAOYSA-N
2-2-chlorophenoxy acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide hydrochloride, 2-2-chlorophenoxy acetimidamide hydrochloride, 2-2-chloro-phenoxy-acetamidine hydrochloride, 2-2-chlorophenoxy ethanimidamide monohydrochloride, 2-2-chlorophenoxy acetamide hydrochloride, ethanimidamide, 2-2-chlorophenoxy-, monohydrochloride, 2-2-chlorophenoxy ethanamidine, chloride, 2-2-chlorophenoxy acetamidine hcl, 2-chlorophenoxy ethanimidamide hydrochloride
2745914
[H+].[Cl-].NC(=N)COC1=CC=CC=C1Cl
Materiale o nome chimico 2-(2-Chlorophenoxy)ethanimidamide hydrochloride
CAS min. % 97.0
Intervallo percentuale saggio 97%
Imballaggio Amber glass bottle
Formula lineare ClC6H4OCH2C(=NH)NH2·HCl
Quantità 1 g
IUPAC Name hydrogen 2-(2-chlorophenoxy)ethanimidamide chloride
Peso formulazione 221.09
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'