missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichloro Diphenyl Disulfide, TRC

Codice prodotto. 30378062
Change view
Click to view available options
Quantità:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30378062 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378062 Supplier Toronto Research Chemicals Supplier No. TRCD434545250MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 2,2'-Dichloro Diphenyl Disulfide
CAS 31121-19-4
Formula molecolare C12H8Cl2S2
Quantità 250 mg
Sinonimo Bis(2-chlorophenyl) Disulfide, Bis(o-chlorophenyl) Disulfide, NSC 32024, 1,2-Bis(2-chlorophenyl)disulfane
InChI Formula InChI=1S/C12H8Cl2S2/c13-9-5-1-3-7-11(9)15-16-12-8-4-2-6-10(12)14/h1-8H
SMILES Clc1ccccc1SSc2ccccc2Cl
Molecular Weight (g/mol) 287.23
Peso formulazione 285.9444
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.