Learn More
2,2'-Dinaphthyl Ether, TRC
Product Code. 30388629
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388629 2.5 g -
30375461 1 g -
30376595 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30388629 Supplier Toronto Research Chemicals Supplier No. TRCD4739202.5G
 Restricted Item
Spedizione stimata: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 613-80-9
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.32
Synonym 2,2'-Oxybis-naphthalene, 2,2'-Oxybis[naphthalene], Di(2-Naphthyl) Ether, Di-beta-Naphthyl Ether
SMILES O(c1ccc2ccccc2c1)c3ccc4ccccc4c3
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C20H14O/c1-3-7-17-13-19(11-9-15(17)5-1)21-20-12-10-16-6-2-4-8-18(16)14-20/h1-14H
Formula Weight 270.1045
Chemical Name or Material 2,2'-Dinaphthyl Ether