Learn More
2,3-Dimethyl-quinoxaline-6-carboxylic acid, 97%, Thermo Scientific™
Product Code. 10707875
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10707875 10 g 10g
10304622 1 g 1g
10609515 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10707875 Supplier Thermo Scientific Maybridge Supplier No. CC27701EA
 Restricted Item
Spedizione stimata: 04-12-2026
Add to basket
Chemical Identifiers
CAS 17635-26-6
Molecular Formula C11H9N2O2
Molecular Weight (g/mol) 201.21
MDL Number MFCD02575516
InChI Key RCACNAWRFYUKLC-UHFFFAOYSA-M
Synonym 2,3-dimethyl-quinoxaline-6-carboxylic acid, 2,3-dimethyl-6-quinoxalinecarboxylic acid, 6-quinoxalinecarboxylicacid, 2,3-dimethyl, 6-quinoxalinecarboxylic acid, 2,3-dimethyl
PubChem CID 236268
SMILES CC1=C(C)N=C2C=C(C=CC2=N1)C([O-])=O
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
IUPAC Name 2,3-dimethylquinoxaline-6-carboxylic acid
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethyl-quinoxaline-6-carboxylic acid