Learn More
2,3-Naphthalic Anhydride, TRC
Product Code. 30481563
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481563 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30481563 Supplier Toronto Research Chemicals Supplier No. TRCN363000100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3-Naphthalic Anhydride
CAS 716-39-2
Molecular Formula C12 H6 O3
Quantity 100 mg
Synonym Naphtho[2,3-c]furan-1,3-dione
InChI Formula InChI=1S/C12H6O3/c13-11-9-5-7-3-1-2-4-8(7)6-10(9)12(14)15-11/h1-6H
SMILES O=C1OC(=O)c2cc3ccccc3cc12
IUPAC Name benzo[f][2]benzofuran-1,3-dione
Molecular Weight (g/mol) 198.17
Formula Weight 198.0317