missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-206461
missing translation for 'orderingAttributeHoverText'
Quantità:
1 g
10 g
25 g
250 g
Imballaggio:
Glass bottle
This item is not returnable. View return policy
830-79-5
C10H12O4
196.2
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
70019
2,4,6-trimethoxybenzaldehyde
COC1=CC(=C(C(=C1)OC)C=O)OC
This item is not returnable. View return policy
830-79-5
C10H12O4
196.2
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
70019
2,4,6-trimethoxybenzaldehyde
COC1=CC(=C(C(=C1)OC)C=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2, 4, 6-Trimethoxybenzaldehyde
Nota nome 98%
CAS min. % 97.5
CAS max % 100.0
Punto di fusione 118.0°C to 120.0°C
Colore Beige
Spettro a infrarossi Authentic
Intervallo percentuale saggio 98%
Imballaggio Glass bottle
Formula lineare (CH3O)3C6H2CHO
Quantità 1 g
Beilstein 08,39
Peso formulazione 196.2
Percent Purity 98%
Forma fisica Crystalline Powder
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'