Learn More
2,4-Bis(1-methylethyl)phenyl Diphenyl Ester Phosphoric Acid, TRC
Codice prodotto. 30342879
Cambia vista
Click to view available options
Quantità:
500 mg
Dimensione della confezione:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30342879 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30342879 Fornitore Toronto Research Chemicals N. del fornitore TRCB451480500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 2,4-Bis(1-methylethyl)phenyl Diphenyl Ester Phosphoric Acid
CAS 96107-55-0
Note sul grado di purezza HPLC
Formula molecolare C24 H27 O4 P
Quantità 500 mg
Sinonimo 2,4-diisopropylphenyl Diphenyl Phosphate
InChI Formula InChI=1S/C24H27O4P/c1-18(2)20-15-16-24(23(17-20)19(3)4)28-29(25,26-21-11-7-5-8-12-21)27-22-13-9-6-10-14-22/h5-19H,1-4H3
SMILES CC(C)c1ccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)c(c1)C(C)C
IUPAC Name (2,4-diisopropylphenyl) diphenyl phosphate
Molecular Weight (g/mol) 410.44
Peso formulazione 410.1647
Percent Purity >95