Learn More
2,4-Dichloro-1,3,5-triazine, TRC
Product Code. 30378823
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378823 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30378823 Supplier Toronto Research Chemicals Supplier No. TRCD4359955G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Dichloro-1,3,5-triazine
CAS 2831-66-5
Molecular Formula C3HCl2N3
Quantity 5 g
Synonym 2,4-Dichloro-s-triazine, 2,4-Dichloro-sym-triazine, 2,4-Dichlorotriazine, 2,6-Dichloro-1,3,5-triazine, 4,6-Dichloro-s-triazine, Dichloro-s-triazine, Eco-Ever 6, Ekoeba 6
InChI Formula InChI=1S/C3HCl2N3/c4-2-6-1-7-3(5)8-2/h1H
SMILES Clc1ncnc(Cl)n1
IUPAC Name 2,4-dichloro-1,3,5-triazine
Molecular Weight (g/mol) 149.97
Formula Weight 148.9548