Learn More
2,5-Bis(4-hydroxyphenyl)piperazine Dihydrochloride, TRC
Codice prodotto. 30329911
Cambia vista
Click to view available options
Quantità:
1 g
Dimensione della confezione:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30329911 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30329911 Fornitore Toronto Research Chemicals N. del fornitore TRCB4473501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 2,5-Bis(4-hydroxyphenyl)piperazine Dihydrochloride
CAS 94572-68-6
Formula molecolare C16H20Cl2N2O2
Quantità 1 g
Sinonimo 4,4'-(2,5-Piperazinediyl)diphenol Dihydrochloride
InChI Formula InChI=1S/C16H18N2O2.2ClH/c19-13-5-1-11(2-6-13)15-9-18-16(10-17-15)12-3-7-14(20)8-4-12;;/h1-8,15-20H,9-10H2;2*1H
SMILES OC(C=C1)=CC=C1C2CNC(C3=CC=C(O)C=C3)CN2
IUPAC Name 4,4'-(piperazine-2,5-diyl)diphenol Dihydrogen chloride
Molecular Weight (g/mol) 343.25
Peso formulazione 342.09