Learn More
2,5-Diformylfuran, TRC
Product Code. 30369606
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30369606 Supplier Toronto Research Chemicals Supplier No. TRCD4459051G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,5-Diformylfuran
CAS 823-82-5
Molecular Formula C6 H4 O3
Quantity 1 g
InChI Formula InChI=1S/C6H4O3/c7-3-5-1-2-6(4-8)9-5/h1-4H
SMILES O=Cc1oc(C=O)cc1
IUPAC Name furan-2,5-dicarbaldehyde
Molecular Weight (g/mol) 124.09
Formula Weight 124.016