Learn More
2,5-Dihydroxybenzaldehyde, TRC
Product Code. 30380307
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380307 25 g -
30372749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30380307 Supplier Toronto Research Chemicals Supplier No. TRCD45100525G
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1194-98-5
Molecular Formula C7 H6 O3
Molecular Weight (g/mol) 138.12
Synonym Benzaldehyde, 2,5-dihydroxy-, Gentisaldehyde (6CI,7CI,8CI), 2,5-Dihydroxybenzaldehyde, 5-Hydroxysalicylaldehyde, Formylhydroquinone, NSC 72387
IUPAC Name 2,5-dihydroxybenzaldehyde
SMILES Oc1ccc(O)c(C=O)c1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C7H6O3/c8-4-5-3-6(9)1-2-7(5)10/h1-4,9-10H
Formula Weight 138.0317
Chemical Name or Material 2,5-Dihydroxybenzaldehyde